MAT047022698 Matrix Scientific

3-Amino-4-(2-methoxyphenoxy)benzotrifluoride

  • MDL number MFCD00052419
  • Cas Number : 175135-08-7
  • Molecular Weight : 283.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 28
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/001317.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001317.html
Availability details for
Quantity
  • Availability
  • Add to Cart