MAT047022706 Matrix Scientific

2,5-Dimethoxyphenylacetonitrile

  • MDL number MFCD00016388
  • Cas Number : 18086-24-3
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-55
Purity 98
Country of Origin USA
Boiling point 167� Torr
Product Link http://www.matrixscientific.com/004019.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004019.html
Availability details for
Quantity
  • Availability
  • Add to Cart