MAT047022727 Matrix Scientific

3,5-Dimethyl-4-iodoisoxazole

  • MDL number MFCD00173743
  • Cas Number : 10557-85-4
  • Molecular Weight : 223.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-54
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/004051.html

Safety Information

hcodes Light Sensitive/Store in Freezer
MSDS http://www.matrixscientific.com/004051.html
Availability details for
Quantity
  • Availability
  • Add to Cart