MAT047022737 Matrix Scientific

2,4-Dimethylbenzonitrile

  • MDL number MFCD00016377
  • Cas Number : 21789-36-6
  • Molecular Weight : 131.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 24-25
Country of Origin USA
Boiling point 112� Torr
Product Link http://www.matrixscientific.com/004062.html

Safety Information

hcodes Irritant-Harmful/Refrigerate
MSDS http://www.matrixscientific.com/004062.html
Availability details for
Quantity
  • Availability
  • Add to Cart