MAT047022748 Matrix Scientific

2,3-Dimethylbenzylamine

  • MDL number MFCD00052680
  • Cas Number : 51586-20-0
  • Molecular Weight : 135.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 0.97
Country of Origin USA
Boiling point 101-103�mm
Product Link http://www.matrixscientific.com/004073.html

Safety Information

hcodes Corrosive/Air Sensitive
MSDS http://www.matrixscientific.com/004073.html
Availability details for
Quantity
  • Availability
  • Add to Cart