MAT047022750 Matrix Scientific

2,5-Dimethylbenzylamine

  • MDL number MFCD00039780
  • Cas Number : 93-48-1
  • Molecular Weight : 135.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 0.95
Country of Origin USA
Boiling point 121�mm
Product Link http://www.matrixscientific.com/004075.html

Safety Information

hcodes Irritant/Air Sensitive
MSDS http://www.matrixscientific.com/004075.html
Availability details for
Quantity
  • Availability
  • Add to Cart