MAT047022752 Matrix Scientific

2,6-Dimethylhydroquinone

  • MDL number MFCD00016466
  • Cas Number : 654-42-2
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-150
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/004077.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/004077.html
Availability details for
Quantity
  • Availability
  • Add to Cart