MAT047022757 Matrix Scientific

2,5-Dimethylphenethylamine

  • MDL number MFCD01708273
  • Cas Number : 23068-44-2
  • Molecular Weight : 149.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 0.94
Country of Origin USA
Boiling point 108�m
Product Link http://www.matrixscientific.com/004081.html

Safety Information

hcodes Corrosive/Irritant-Harmful
MSDS http://www.matrixscientific.com/004081.html
Availability details for
Quantity
  • Availability
  • Add to Cart