MAT047022759 Matrix Scientific

2,5-Dimethylphenyl isothiocyanate

  • MDL number MFCD00046798
  • Cas Number : 19241-15-7
  • Molecular Weight : 163.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.08
Country of Origin USA
Boiling point 262
Product Link http://www.matrixscientific.com/004084.html

Safety Information

hcodes Toxic/Corrosive
MSDS http://www.matrixscientific.com/004084.html
Availability details for
Quantity
  • Availability
  • Add to Cart