MAT047022767 Matrix Scientific

2,4-Dimethylthiazole

  • MDL number MFCD00014509
  • Cas Number : 541-58-2
  • Molecular Weight : 113.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-71
Purity 97
Density 1.057
Country of Origin USA
Boiling point 145-147
Product Link http://www.matrixscientific.com/004097.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004097.html
Availability details for
Quantity
  • Availability
  • Add to Cart