MAT047022856 Matrix Scientific

4-Fluorodiphenyl ether

  • MDL number MFCD00055239
  • Cas Number : 330-84-7
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 1.1523/16
Country of Origin USA
Boiling point 254-255�0 Torr
Product Link http://www.matrixscientific.com/004219.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004219.html
Availability details for
Quantity
  • Availability
  • Add to Cart