MAT047022862 Matrix Scientific

2-Fluorodiphenylmethane

  • MDL number MFCD00079758
  • Cas Number : 3794-15-8
  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.08
Country of Origin USA
Boiling point 74�2 Torr
Product Link http://www.matrixscientific.com/004225.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004225.html
Availability details for
Quantity
  • Availability
  • Add to Cart