MAT047022873 Matrix Scientific

Fluoromesitylene

  • MDL number MFCD00052365
  • Cas Number : 392-69-8
  • Molecular Weight : 138.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -36
Purity 99
Density 0.9745/25
Storage temp. Ambient
Country of Origin USA
Boiling point 167�4 Torr
Product Link http://www.matrixscientific.com/004246.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004246.html
Availability details for
Quantity
  • Availability
  • Add to Cart