MAT047022904 Matrix Scientific

3,4-Methylenedioxybenzyl chloride (50% by weight in methylene chloride)

  • MDL number MFCD00044328
  • Cas Number : 20850-43-5
  • Molecular Weight : 170.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 20
Purity 98
Country of Origin USA
Boiling point 134-135� Torr
Product Link http://www.matrixscientific.com/004299.html

Safety Information

hcodes Irritant/Moisture Sensitive/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/004299.html
Availability details for
Quantity
  • Availability
  • Add to Cart