MAT047022917 Matrix Scientific

4-Cyano-3-(trifluoromethyl)acetanilide

  • MDL number MFCD00052360
  • Cas Number : 97760-99-1
  • Molecular Weight : 228.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-173
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/001346.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001346.html
Availability details for
Quantity
  • Availability
  • Add to Cart