MAT047022939 Matrix Scientific

2-Nitro-4-(trifluoromethoxy)acetanilide

  • MDL number MFCD00052324
  • Cas Number : 787-57-5
  • Molecular Weight : 264.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-99
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/001348.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001348.html
Availability details for
Quantity
  • Availability
  • Add to Cart