MAT047022950 Matrix Scientific

2-Nitro-4-(trifluoromethoxy)aniline

  • MDL number MFCD00042326
  • Cas Number : 2267-23-4
  • Molecular Weight : 222.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-64
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001349.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/001349.html
Availability details for
Quantity
  • Availability
  • Add to Cart