MAT047022958 Matrix Scientific

3H-Heptafluoro-2,2,4,4-tetrahydroxypentane

  • MDL number MFCD02093308
  • Cas Number : 77953-71-0
  • Molecular Weight : 262.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-114
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/004373.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004373.html
Availability details for
Quantity
  • Availability
  • Add to Cart