MAT047022972 Matrix Scientific

4-Nitro-3-(trifluoromethyl)acetanilide

  • MDL number MFCD00017994
  • Cas Number : 393-12-4
  • Molecular Weight : 248.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-109
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001351.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001351.html
Availability details for
Quantity
  • Availability
  • Add to Cart