MAT047023005 Matrix Scientific

3-Bromo-4-(trifluoromethoxy)aniline

  • MDL number MFCD00190127
  • Cas Number : 191602-54-7
  • Molecular Weight : 256.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001354.html

Safety Information

hcodes Toxic/Store in Freezer
MSDS http://www.matrixscientific.com/001354.html
Availability details for
Quantity
  • Availability
  • Add to Cart