MAT047023008 Matrix Scientific

2',3'-Difluoropropiophenone

  • MDL number MFCD00061141
  • Cas Number : 236754-63-5
  • Molecular Weight : 170.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/004429.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004429.html
Availability details for
Quantity
  • Availability
  • Add to Cart