MAT047023017 Matrix Scientific

2',6'-Difluoropropiophenone

  • MDL number MFCD00015506
  • Cas Number : 85068-31-1
  • Molecular Weight : 170.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 71�m
Product Link http://www.matrixscientific.com/004437.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004437.html
Availability details for
Quantity
  • Availability
  • Add to Cart