MAT047023039 Matrix Scientific

2,3,4-Trimethoxyphenylacetic acid

  • MDL number MFCD00016820
  • Cas Number : 22480-91-7
  • Molecular Weight : 226.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/004466.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004466.html
Availability details for
Quantity
  • Availability
  • Add to Cart