MAT047023062 Matrix Scientific

3-Iodo-2-methylbenzoic acid

  • MDL number MFCD00270112
  • Cas Number : 133232-56-1
  • Molecular Weight : 262.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-126
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/004490.html

Safety Information

hcodes Light Sensitive
MSDS http://www.matrixscientific.com/004490.html
Availability details for
Quantity
  • Availability
  • Add to Cart