MAT047023076 Matrix Scientific

3-(Hexylamino)propionitrile

  • MDL number MFCD00060315
  • Cas Number : 55490-85-2
  • Molecular Weight : 154.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 0.86
Country of Origin USA
Boiling point 130-134� Torr
Product Link http://www.matrixscientific.com/004511.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/004511.html
Availability details for
Quantity
  • Availability
  • Add to Cart