MAT047023083 Matrix Scientific

Ethyl 2-amino-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD00153147
  • Cas Number : 344-72-9
  • Molecular Weight : 240.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-171
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/004517.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004517.html
Availability details for
Quantity
  • Availability
  • Add to Cart