MAT047023091 Matrix Scientific

1,2-Dibromo-1,1-dichlorodifluoroethane

  • MDL number MFCD00039319
  • Cas Number : 558-57-6
  • Molecular Weight : 292.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46
Purity 97
Country of Origin USA
Boiling point 138-140
Product Link http://www.matrixscientific.com/004530.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/004530.html
Availability details for
Quantity
  • Availability
  • Add to Cart