MAT047023099 Matrix Scientific

2,3-Dibromo-2-(trifluoromethyl)-1,1,1-trifluoro-propane

  • MDL number MFCD01862098
  • Cas Number : 247220-90-2
  • Molecular Weight : 323.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 115-116
Product Link http://www.matrixscientific.com/004537.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004537.html
Availability details for
Quantity
  • Availability
  • Add to Cart