MAT047023101 Matrix Scientific

2,3-Dibromo-2-chloro-1,1,1,4,4,4-hexafluorobutane

  • MDL number MFCD01862049
  • Cas Number : 63573-66-0
  • Molecular Weight : 358.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 78�mm
Product Link http://www.matrixscientific.com/004539.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004539.html
Availability details for
Quantity
  • Availability
  • Add to Cart