MAT047023141 Matrix Scientific

2,3-Dibromohexafluoro-2-butene

  • MDL number MFCD01862090
  • Cas Number : 384-51-0
  • Molecular Weight : 321.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 106
Product Link http://www.matrixscientific.com/004579.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004579.html
Availability details for
Quantity
  • Availability
  • Add to Cart