MAT047023149 Matrix Scientific

2,5-Dibromophenyl isothiocyanate

  • MDL number MFCD00041041
  • Cas Number : 98041-67-9
  • Molecular Weight : 292.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49-51
Purity 98
Storage temp. Refrigerate
Country of Origin USA
Product Link http://www.matrixscientific.com/004587.html

Safety Information

hcodes Irritant/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/004587.html
Availability details for
Quantity
  • Availability
  • Add to Cart