MAT047023194 Matrix Scientific

2-Iodo-4-nitroanisole

  • MDL number MFCD00024328
  • Cas Number : 5399-03-1
  • Molecular Weight : 279.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94
Purity 99
Country of Origin USA
Product Link http://www.matrixscientific.com/004650.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004650.html
Availability details for
Quantity
  • Availability
  • Add to Cart