MAT047023196 Matrix Scientific

2-Iodo-5-nitroanisole

  • MDL number MFCD00024590
  • Cas Number : 5458-84-4
  • Molecular Weight : 279.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-135
Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/004651.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004651.html
Availability details for
Quantity
  • Availability
  • Add to Cart