MAT047023208 Matrix Scientific

1,2-Diamino-3,4,5,6-tetrafluorobenzene

  • MDL number MFCD00278787
  • Cas Number : 2993-07-9
  • Molecular Weight : 180.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-132
Country of Origin USA
Product Link http://www.matrixscientific.com/004665.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/004665.html
Availability details for
Quantity
  • Availability
  • Add to Cart