MAT047023211 Matrix Scientific

2,3-Diamino-6-(trifluoromethyl)-4(3H)-pyrimidinone

  • MDL number MFCD00068010
  • Cas Number : 95095-71-9
  • Molecular Weight : 194.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217-219
Country of Origin USA
Product Link http://www.matrixscientific.com/004669.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004669.html
Availability details for
Quantity
  • Availability
  • Add to Cart