MAT047023212 Matrix Scientific

1,2-Diamino-4-chloro-5-fluorobenzene

  • MDL number MFCD00042485
  • Cas Number : 139512-70-2
  • Molecular Weight : 160.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/004670.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004670.html
Availability details for
Quantity
  • Availability
  • Add to Cart