MAT047023214 Matrix Scientific

5-Fluoro-2,4-quinazolinediamine

  • MDL number MFCD00221785
  • Cas Number : 119584-70-2
  • Molecular Weight : 178.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249-251
Country of Origin USA
Product Link http://www.matrixscientific.com/004672.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004672.html
Availability details for
Quantity
  • Availability
  • Add to Cart