MAT047023222 Matrix Scientific

9-Hydroxynonanoic acid

  • MDL number MFCD00792445
  • Cas Number : 3788-56-5
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-54
Country of Origin USA
Product Link http://www.matrixscientific.com/004687.html

Safety Information

MSDS http://www.matrixscientific.com/004687.html
Availability details for
Quantity
  • Availability
  • Add to Cart