MAT047023256 Matrix Scientific

4-Bromobenzhydrol

  • MDL number MFCD00016851
  • Cas Number : 29334-16-5
  • Molecular Weight : 263.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/004731.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004731.html
Availability details for
Quantity
  • Availability
  • Add to Cart