MAT047023321 Matrix Scientific

(Perfluoro-n-octyl)phenyliodoniumtrifluoromethanesulfonate

  • MDL number MFCD00135120
  • Cas Number : 77758-89-5
  • Molecular Weight : 772.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/004811.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004811.html
Availability details for
Quantity
  • Availability
  • Add to Cart