MAT047023322 Matrix Scientific

(Perfluoro-n-octyl)ethane

  • MDL number MFCD00236711
  • Cas Number : 77117-48-7
  • Molecular Weight : 448.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -38
Purity 97
Density 1.654
Country of Origin USA
Boiling point 155
Product Link http://www.matrixscientific.com/004812.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004812.html
Availability details for
Quantity
  • Availability
  • Add to Cart