MAT047023329 Matrix Scientific

Hexafluoroacetone sesquihydrate

  • MDL number MFCD06800792
  • Cas Number : 13098-39-0
  • Molecular Weight : 386.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 18-21
Purity 98
Density 1.69
Country of Origin USA
Product Link http://www.matrixscientific.com/004820.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/004820.html
Availability details for
Quantity
  • Availability
  • Add to Cart