MAT047023336 Matrix Scientific

Perfluoro-15-crown-5-ether

  • MDL number MFCD01320747
  • Cas Number : 97571-69-2
  • Molecular Weight : 580.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 1.78
Storage temp. Ambient
Country of Origin USA
Boiling point 145
Product Link http://www.matrixscientific.com/001399.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001399.html
Availability details for
Quantity
  • Availability
  • Add to Cart