MAT047023361 Matrix Scientific

Perfluorooctanonitrile

  • MDL number MFCD00039481
  • Cas Number : 647-12-1
  • Molecular Weight : 395.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 103-104
Product Link http://www.matrixscientific.com/004855.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/004855.html
Availability details for
Quantity
  • Availability
  • Add to Cart