MAT047023379 Matrix Scientific

2,5-Diiodofluorobenzene

  • MDL number MFCD00061107
  • Cas Number : 147808-02-4
  • Molecular Weight : 347.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-71
Purity 99
Country of Origin USA
Boiling point 135�mm
Product Link http://www.matrixscientific.com/004876.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004876.html
Availability details for
Quantity
  • Availability
  • Add to Cart