MAT047023404 Matrix Scientific

1-(4-Chloro-2-fluoro-5-methylphenyl)-1-ethanone

  • MDL number MFCD00236601
  • Cas Number : 177211-26-6
  • Molecular Weight : 186.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/001406.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001406.html
Availability details for
Quantity
  • Availability
  • Add to Cart