MAT047023419 Matrix Scientific

4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile

  • MDL number MFCD00173780
  • Cas Number : 175277-01-7
  • Molecular Weight : 264.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-85
Country of Origin USA
Product Link http://www.matrixscientific.com/004925.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004925.html
Availability details for
Quantity
  • Availability
  • Add to Cart