MAT047023421 Matrix Scientific

4-(3-Bromophenyl)-3-pyridinecarboxaldehyde

  • MDL number MFCD01310796
  • Cas Number : 376646-64-9
  • Molecular Weight : 262.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-86
Storage temp. argon
Country of Origin USA
Product Link http://www.matrixscientific.com/004927.html

Safety Information

hcodes Irritant/Air Sensitive/Stored under Argon
MSDS http://www.matrixscientific.com/004927.html
Availability details for
Quantity
  • Availability
  • Add to Cart