MAT047023422 Matrix Scientific

1-(4-Nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carbonyl chloride

  • MDL number MFCD00173866
  • Cas Number : 175137-36-7
  • Molecular Weight : 319.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-79
Country of Origin USA
Product Link http://www.matrixscientific.com/004929.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/004929.html
Availability details for
Quantity
  • Availability
  • Add to Cart