MAT047023435 Matrix Scientific

3-(3-Bromophenyl)propionitrile

  • MDL number MFCD01310794
  • Cas Number : 376646-63-8
  • Molecular Weight : 210.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.42
Country of Origin USA
Boiling point 113�m
Product Link http://www.matrixscientific.com/004942.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/004942.html
Availability details for
Quantity
  • Availability
  • Add to Cart